Learn More
Rat IgG2a kappa Isotype Control (eBR2a), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Isotype Control
174.00 CHF - 371.00 CHF
Spezifikation
Antigen | Rat IgG2a kappa |
---|---|
Klon | eBR2a |
Konzentration | 0.2 mg/mL |
Anwendungen | Control, Flow Cytometry |
Konjugat | Super Bright 436 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15805533
|
Affymetrix eBioscience
62-4321-80 |
25 μg |
174.00 CHF
25 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
16119920
|
Affymetrix eBioscience
62-4321-82 |
100 μg |
371.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin. Applications Reported: This eBR2a antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBR2a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Rat IgG2a kappa | |
0.2 mg/mL | |
Super Bright 436 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.