missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa Isotype Control (eBRG1), Super Bright™ 702, eBioscience™, Invitrogen™

Rat Isotype Control

178.00 CHF - 365.00 CHF

Spezifikation

Antigen Rat IgG1 kappa
Klon eBRG1
Konzentration 0.2 mg/mL
Anwendungen Control, Flow Cytometry
Konjugat Super Bright 702
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15983754
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
67-4301-80
25 μg
178.00 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15993754
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
67-4301-82
100 μg
365.00 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in flow cytometric analysis and ELISA. Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension, This can be used at the same concentration as the experimental antibody. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Spezifikation

Spezifikation

Rat IgG1 kappa
0.2 mg/mL
Super Bright 702
Rat
Not Applicable
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
eBRG1
Control, Flow Cytometry
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
IgG1 κ
Affinity chromatography
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications