Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Isotype Control
Marke: Affymetrix eBioscience 62-4301-82
Additional Details : Gewicht : 0.01000kg
Beschreibung
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA. Applications Tested: Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.Spezifikation
Rat IgG1 kappa | |
eBRG1 | |
Super Bright 436 | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Rat | |
100μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |