missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA565036
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: ILGGRDSHPAA GGGSVLCFGQ CQYTAEEYQA IQKALRQRLG PEYISSRMAG GGQKVCYIEG HRVINLANEM FGYNGWAHSI TQQNVDFVDL NNGKFYVGVC AFVRVQLKDG SYHEDVGYGV SE Highest antigen sequence identity to the following orthologs - mouse 90%, rat 90%.
DNA double-strand breaks can be generated by exposure to ionizing radiation or through endogenous cellular reactions. Double-strand breaks are generally thought to be repaired via one of two distinct mechanisms: homologous recombination (HR) or non-homologous end joining (NHEJ). Homologous recombination requires the activity of the members of the RAD52 epistasis group, which includeRAD50, RAD51, RAD52, RAD54, RDH54/TID1, RAD55, RAD57, RAD59, MRE11, and XRS2. RAD52 can catalyze strand annealing reactions, and can form oligomers that attach to ends of single-stranded DNA.
Spezifikation
| RAD52 | |
| Polyclonal | |
| Unconjugated | |
| RAD52 | |
| DNA repair protein RAD52 homolog; RAD52; RAD52 homolog (S. cerevisiae); RAD52 homolog, DNA repair protein; Rad52yh; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43351 | |
| RAD52 | |
| Recombinant Human RAD52. Recombinant protein control fragment (Product #RP-105801). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur