missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RAC2 Recombinant Protein
Marke: Abnova™ H00005880-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH01485.1 | |
Solution | |
5880 | |
RAC2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAC2 | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
46.86 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MQAIKCVVVGDGAVGKTCLLISYTTNAFPGEYIPTVFDNYSANVMVDSKPVNLGLWDTAGQEDYDRLRPLSYPQTDVFLICFSLVSPASYENVRAKWFPEVRHHCPSTPIILVGTKLDLRDDKDTIEKLKEKKLAPITYPQGLALAKEIDSVKYLECSALTQRGLKTVFDEAIRAVLCPQPTRQQKRACSLL | |
EN-7/Gx/HSPC022 | |
RAC2 | |
Wheat Germ (in vitro) | |
GST |