missing translation for 'onlineSavingsMsg'
Learn More

rac-Pinocembrin, TRC

Product Code. 30240709
Change view
Click to view available options
Menge:
250 mg
5 g
Unit Size:
250 Milligramm
5 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30240709 5 g 5 Gramm
30240612 250 mg 250 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240709 Supplier Toronto Research Chemicals Supplier No. TRCP4684955G
 Eingeschränkter Artikel
Verfügbar ab: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68745-38-0
Summenformel C15 H12 O4
Molekulargewicht (g/mol) 256.25
Synonym Flavanone, 5,7-dihydroxy- (7CI), 2,3-Dihydro-5,7-dihydroxy-2-phenyl-4H-1-benzopyran-4-one, (±)-5,7-Dihydroxyflavanone, (±)-Pinocembrin, NSC 43318, rac-Pinocembrin
IUPAC-Name 5,7-Dihydroxy-2-Phenyl-2,3-Dihydrochromen-4-On
SMILES Oc1cc(O)c2C(=O)CC(Oc2c1)c3ccccc3

Specifications

Chemischer Name oder Material Pinocembrin
Menge 5 g
InChI-Formel InChI=1S/C15H12O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-7,13,16-17H,8H2
Formelmasse 256.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.