Learn More
rac-erythro-Dihydro Bupropion Hydrochloride, TRC
Artikelnummer. 30132702
Change view
Click to view available options
Menge:
100 mg
Packungsgröße:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30132702 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30132702 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD448730100MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 80478-43-9
Summenformel C13 H20 Cl N O . HCl
Molekulargewicht (g/mol) 278.22
Synonym rac erythro-2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-ol Hydrochloride
SMILES ClC1=CC([C@@H](O)[C@H](C)NC(C)(C)C)=CC=C1.Cl
Spezifikation
Chemischer Name oder Material (±)-erythro-Dihydrobupropion Hydrochloride
Menge 100 mg
InChI-Formel InChI=1S/C13H20ClNO/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10/h5-9,12,15-16H,1-4H3/t9-,12-/m1/s1
Formelmasse 277.1