Learn More
rac-cis-2,3-Dichloro Sertraline Hydrochloride, TRC
Artikelnummer. 30009946
Change view
Click to view available options
Menge:
5 mg
10 mg
100 mg
Packungsgröße:
10 Milligramm
100 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30009946 100 mg 100 Milligramm
30006238 10 mg 10 Milligramm
30009002 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009946 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD435880100MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1198084-29-5
Summenformel C17 H17 Cl2 N . Cl H
Molekulargewicht (g/mol) 342.6905
Synonym 1-Naphthalenamine, 4-(2,3-dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-, hydrochloride (1:1), (1S,4R)-, (1S,4R)-4-(2,3-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride (1:1), cis-2,3-Dichloro sertraline hydrochloride, cis-2,3-Dichlorosertraline hydrochloride, (1S-cis)-2,3-Dichlorosertraline hydrochloride, (1S,4R)-4-(2,3-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methylnaphthalenamine hydrochloride, (1S,4R)-4-(2,3-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
IUPAC-Name (1S,4R)-4-(2,3-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES Cl.CN[C@H]1CC[C@@H](c2cccc(Cl)c2Cl)c3ccccc13
Spezifikation
Chemischer Name oder Material rac-cis-2,3-Dichloro Sertraline Hydrochloride
Hinweise zur Reinheitsqualität HPLC
Menge 100 mg
InChI-Formel InChI=1S/C17H17Cl2N.ClH/c1-20-16-10-9-12(11-5-2-3-6-13(11)16)14-7-4-8-15(18)17(14)19;/h2-8,12,16,20H,9-10H2,1H3;1H/t12-,16+;/m1./s1
Formelmasse 341.0505
Reinheit (%) >95%