missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-2,3-Dichloro Sertraline Hydrochloride, TRC

Artikelnummer. 30006238
Change view
Click to view available options
Menge:
5 mg
10 mg
100 mg
Packungsgröße:
10 Milligramm
100 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30006238 10 mg 10 Milligramm
30009002 5 mg 5 Milligramm
30009946 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30006238 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD43588010MG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1198084-29-5
Summenformel C17 H17 Cl2 N . Cl H
Molekulargewicht (g/mol) 342.6905
Synonym 1-Naphthalenamine, 4-(2,3-dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-, hydrochloride (1:1), (1S,4R)-, (1S,4R)-4-(2,3-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride (1:1), cis-2,3-Dichloro sertraline hydrochloride, cis-2,3-Dichlorosertraline hydrochloride, (1S-cis)-2,3-Dichlorosertraline hydrochloride, (1S,4R)-4-(2,3-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methylnaphthalenamine hydrochloride, (1S,4R)-4-(2,3-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
IUPAC-Name (1S,4R)-4-(2,3-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES Cl.CN[C@H]1CC[C@@H](c2cccc(Cl)c2Cl)c3ccccc13

Spezifikation

Chemischer Name oder Material rac-cis-2,3-Dichloro Sertraline Hydrochloride
Hinweise zur Reinheitsqualität HPLC
Menge 10 mg
InChI-Formel InChI=1S/C17H17Cl2N.ClH/c1-20-16-10-9-12(11-5-2-3-6-13(11)16)14-7-4-8-15(18)17(14)19;/h2-8,12,16,20H,9-10H2,1H3;1H/t12-,16+;/m1./s1
Formelmasse 341.0505
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.