Learn More
ACROBiosystems Rabbit TIGIT Protein, Fc Tag
Marke: ACROBiosystems TIT-R5259-50UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
T-cell immunoreceptor with Ig and ITIM domains (TIGIT) is also known as V-set and immunoglobulin domain-containing protein 9 (VSIG9), V-set and transmembrane domain-containing protein 3 (VSTM3),which belongs to single-pass type I membrane protein containing an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells (at protein level). TIGIT binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. As a result of glycosylation, the protein migrates as 45-50 kDa under reducing (R) condition, and 90-116 kDa under non-reducing (NR) condition (SDS-PASpezifikation
40.4 kDa | |
Rabbit TIGIT, Fc Tag (TIT-R5259) is expressed from human 293 cells (HEK293). It contains AA Ala 16 - Pro 142 (Accession # G1SLC1). | |
-20°C | |
TIGIT | |
Rabbit | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
50 μg | |
Ala 16 - Pro 142 | |
TIGIT,VSIG9,VSTM3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |