Learn More
CLOCK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CLOCK.
Marke: Abnova PAB2981.400uL
Additional Details : Gewicht : 0.02000kg
Beschreibung
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1. [provided by RefSeq
Spezifikation
CLOCK | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CLOCK. | |
In PBS (0.09% sodium azide) | |
NP_004889;O15516 | |
CLOCK | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human CLOCK. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
CLOCK | |
KAT13D/KIAA0334/bHLHe8 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
9575 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only