Learn More
CDH2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDH2.
Marke: Abnova PAB7892.100uL
Beschreibung
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry. At certain central nervous system synapses, presynaptic to postsynaptic adhesion is mediated at least in part by this gene product. [provided by RefSeq
Spezifikation
| CDH2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDH2. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| P19022 | |
| CDH2 | |
| A synthetic peptide corresponding to amino acids 853-864 of human CDH2. | |
| RUO | |
| 1000 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| CDH2 | |
| CD325/CDHN/CDw325/NCAD | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This sequence is conserved in rat and mouse N-cadherin, as well as in E-, P-, and R-cadherin from human, rat, and mouse. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only