missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casomorphin-beta Rabbit anti-Bovine, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Casomorphin-beta.
Marke: Abnova PAB5091.150ug
Additional Details : Gewicht : 0.02000kg
Beschreibung
Sequence: CYPFPGPISpezifikation
Casomorphin-beta | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Casomorphin-beta. | |
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
MGC133765 | |
Rabbit | |
150 μg | |
Primary | |
Bovine | |
Liquid |
ELISA, Immunoprecipitation | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CSN2 | |
CSN2 | |
A synthetic peptide (conjugated with KLH) corresponding to bovine Casomorphin-beta. | |
RUO | |
281099 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only