missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB8B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | RAB8B |
---|---|
Konzentration | 0.26 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839103
|
Proteintech
11792-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829103
|
Proteintech
11792-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab superfamilies exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. Rab 8B (Ras-related protein Rab-8B) is a 207 amino acid lipid anchor that localizes to the cytoplasmic side of the cell membrane and belongs to the Rab family and small GTPase superfamily. Rab 8B may be involved in neurotransmitter release and vesicular trafficking, and is encoded by a gene located on human chromosome Rab 8B and mouse chromosome 9 C.Spécification
RAB8B | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P61028, Q92930 | |
235442, 51762 | |
RAB8B Fusion Protein Ag2421 | |
Primary | |
-20°C | |
RAB8B |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RAB8, RAB8B, Ras related protein Rab 8B | |
RAB8B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |