missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB43 Mouse anti-Human, Clone: 3B2A8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66654-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. The low intrinsic GTPase activity of RAB43 is activated by USP6NL. Involved in retrograde transport from the endocytic pathway to the Golgi apparatus. Involved in the transport of Shiga toxin from early and recycling endosomes to the trans-Golgi network. Required for the structural integrity of the Golgi complex. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.Spezifikation
RAB43 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86YS6 | |
RAB43 | |
RAB43 Fusion Protein Ag26911 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3B2A8 | |
Unconjugated | |
RAB43 | |
ISY1, RAB11B, RAB41, RAB43, Ras related protein Rab 41, Ras related protein Rab 43 | |
Mouse | |
Protein G | |
RUO | |
339122 | |
-20°C | |
Liquid |