missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19742-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to RAB11B. The antibody has no cross reaction to RAB11A.
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 11 is a known marker for protein trafficking, sorting, and recycling in the endosomal pathway.Spezifikation
RAB11B | |
Polyclonal | |
Unconjugated | |
RAB11B | |
GTP binding protein YPT3, H YPT3, RAB11B, Ras related protein Rab 11B, YPT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19326, 79434, 9230 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35509, P46638, Q15907 | |
RAB11B | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |