Learn More
R&D Systems™ Rat ADAMTS4 (NP_076449) VersaClone cDNA
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
Beschreibung
The Rat ADAMTS4 (NP_076449) VersaClone cDNA clone contains the complete ORF for ADAMTS4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ADAMTS4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature
Spezifikation
Spezifikation
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Menge | 10 μg |
For Reasearch Use Only; Not for Use in Humans