Learn More
R&D Systems™ Human SLCO1C1/OATP1C1 (NP_001139418) VersaClone cDNA
Artikelnummer. 18788089
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18788089 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18788089 Lieferant R&D Systems™ Lieferanten-Nr. RDC1148
  • ONLINE EXCLUSIVE PRICE
    807.00 CHF
    763.35 CHF / 10 Mikrogramm
    Sparen 43.65 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 21-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Solute Carrier Organic Anion Transporter Family, Member 1C1
The Human SLCO1C1/OATP1C1 (NP_001139418) VersaClone cDNA clone contains the complete ORF for SLCO1C1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLCO1C1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Spezifikation
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans