Learn More
R&D Systems™ TACS XTT Cell Proliferation/Viability Assay
Click to view available options
Menge:
2.500 Tests
Packungsgröße:
2500 Tests
Beschreibung
Includes
XTT Reagent, 5 x 25mL; XTT Activator, 5 x 0.5mL
XTT is added directly to the culture medium and cleavage of the tetrazolium salt to formazan occurs in metabolically active cells
The orange formazan dye generated can be measured by absorbance at 490nm (or 450nm) in a microplate reader to yield rapid re
Spezifikation
Spezifikation
| Menge | 2.500 Tests |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |