Learn More
Tocris Bioscience™ IPTG
Used in cloning procedures with X-GAL
Marke: Tocris Bioscience™ 5509/10G
Beschreibung
IPTG is an used in combination with X-GAL (Cat. No. 5510) in cloning procedures requiring expression of β-galactosidase activity. Non-metabolizable analog of galactose.This product has <0.1% dioxane content.
Spezifikation
| 1-Methylethyl 1-thio-β-D-galactopyranoside | |
| 1-Methylethyl 1-thio-β-D-galactopyranoside | |
| Soluble to 100mM in water | |
| Bioactive Small Molecule |
| 367-93-1 | |
| 10 g | |
| Solid |
Research Use Only. Not For Human or Veterinary Use