Learn More
R&D Systems™ Human VSIG8 (NP_001013683) VersaClone cDNA
Beschreibung
The Human VSIG8 (NP_001013683) VersaClone cDNA clone contains the complete ORF for VSIG8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VSIG8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
Spezifikation
Spezifikation
| Produkttyp | cDNA |
| Beschreibung | Gene ID (Entrez): 391123, 240916, 289236 |
| Gen | VSIG8 |
| Zugriffsnummer | NP_001013683 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Einsatztyp | 1258bp |
| Gen-ID (Entrez) | 391123 |
| Gen-Alias | V-set and immunoglobulin domain containing 8 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Mehr anzeigen |
For Research Use Only; Not for Use in Humans