Learn More
R&D Systems™ Human PAR4 (AAC25699) VersaClone cDNA
Artikelnummer. 18451440
Change view
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18451440 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18451440 Lieferant R&D Systems™ Lieferanten-Nr. RDC0170
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Protease-Activated Receptor 4
The Human PAR4 (AAC25699) VersaClone cDNA clone contains the complete ORF for PAR4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PAR4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Gen hPAR4
Zugriffsnummer AAC25699
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10mM Tris-Cl, pH 8.5
Einsatztyp 1170bp
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Human
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans