Learn More
R&D Systems™ Human Leukotriene B4 R1 (NP_858043) VersaClone cDNA
Artikelnummer. 18461620
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18461620 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18461620 Lieferant R&D Systems™ Lieferanten-Nr. RDC0413
  • ONLINE EXCLUSIVE PRICE
    345.00 CHF
    325.50 CHF / 10 Mikrogramm
    Sparen 19.50 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 28-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Leukotriene B4 Receptor
The Human Leukotriene B4 R1 (NP_858043) VersaClone cDNA clone contains the complete ORF for Leukotriene B4 R1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Leukotriene B4 R1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Gen hLTB4R
Zugriffsnummer NP_858043
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10mM Tris-Cl, pH 8.5
Einsatztyp 1072bp
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Human
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans