Learn More
R&D Systems™ Human Integrin alpha 3/CD49c (NP_002195) VersaClone cDNA
Artikelnummer. 18769607
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18769607 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18769607 Lieferant R&D Systems™ Lieferanten-Nr. RDC0403
  • ONLINE EXCLUSIVE PRICE
    807.00 CHF
    763.35 CHF / 10 Mikrogramm
    Sparen 43.65 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 04-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
ITGA3
The Human Integrin alpha 3/CD49c (NP_002195) VersaClone cDNA clone contains the complete ORF for Integrin alpha 3/CD49c, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin alpha 3/CD49c insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Gen hITGA3
Zugriffsnummer NP_002195
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10mM Tris-Cl, pH 8.5
Einsatztyp 3169bp
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Human
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans