Learn More
R&D Systems™ Human ABCA1 (NP_005493) VersaClone cDNA
Artikelnummer. 18471960
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18471960 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18471960 Lieferant R&D Systems™ Lieferanten-Nr. RDC0807
  • ONLINE EXCLUSIVE PRICE
    1260.00 CHF
    1190.70 CHF / 10 Mikrogramm
    Sparen 69.30 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 20-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
ATP-binding Cassette, Sub-family A (ABC1), Member 1
The Human ABCA1 (NP_005493) VersaClone cDNA clone contains the complete ORF for ABCA1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ABCA1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Gen hABCA1
Zugriffsnummer NP_005493
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10mM Tris-Cl, pH 8.5
Einsatztyp 6798bp
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Human
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans