Learn More
Tocris Bioscience™ Aripiprazole
High affinity D2 and 5-HT1A receptor partial agonist; also 5-HT2A antagonist
Brand: Tocris Bioscience™ 5584/50
Description
Aripiprazole is a high affinity dopamine D2 and 5-HT1A receptor partial agonist (Ki values are ∽1.64 and 5.59 nM respectively). Also 5-HT2A antagonist (Ki = 8.7 nM). Suppresses morphine-induced emesis in vivo. Atypical antipsychotic.Specifications
| Aripiprazole | |
| 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 129722-12-9 | |
| 50 mg | |
| Solid |
Research Use Only. Not For Human or Veterinary Use