Learn More
Quickfit™ Halterung für Rührstäbe aus Glas/PTFE
Marke: Quickfit 19146
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Designed to accept SVL™ stirrer shafts with ground glass surfaces
- Borosilicate glass complete with fittings
- Glands manufactured from a PTFE based polymer which is self lubricating and corrosion resistant
- Two types available: - With SVL™ screwthread to allow connection to SVL™ flasks - With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks