missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QA1b Antibody (6A8.6F10.1A6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-26667-0.1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
QA1b Monoclonal antibody specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| QA1b | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1 μL/1 million cells, CyTOF-ready | |
| H2-Q1, H2-T23 | |
| Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells.The antibody reacts with the Qa-1b Class Ib molecule. | |
| 0.1 mg | |
| Signal Transduction | |
| 15040 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, CyTOF | |
| 6A8.6F10.1A6 | |
| Unconjugated | |
| Sterile - filtered PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction