Learn More
Invitrogen™ Qa-2 Monoclonal Antibody (69H1-9-9), FITC, eBioscience™

Mouse Monoclonal Antibody
203.00 CHF
Spezifikation
| Antigen | Qa-2 |
|---|---|
| Klon | 69H1-9-9 |
| Konzentration | 0.5 mg/mL |
| Inhalt und Lagerung | 4°C, store in dark, DO NOT FREEZE! |
| Anwendungen | Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15278329
![]() |
Invitrogen™
11-5996-82 |
100 μg |
203.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Description: The 69H1-9-9 monoclonal antibody reacts with the mouse Qa-2 molecule, a GPI-linked MHC class Ibeta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level is different depending on the laboratory mouse strain. Applications Reported: The 69H1-9-9 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 69H1-9-9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Qa-2 is a GPI-linked MHC class I beta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level varies between laboratory mouse strains.Spezifikation
| Qa-2 | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 0 | |
| 111458 | |
| Primary | |
| Affinity chromatography |
| 69H1-9-9 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Mouse | |
| Qa2 | |
| H2Q7; H2-Qa2; Ped; Qa lymphocyte antigen 2 region; Qa2; Qa-2; Qa-2 antigen | |
| Qa2 | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.