Learn More
Euro Protection™ Purofort™ Safety Boots
Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.
Marke: Euro Protection™ 9PUSG41
Additional Details : Gewicht : 0.01000kg
Beschreibung
- Flexible and lightweight for use all day long
- Cold insulation
- Tear and wear resistant
- Purofort™ upper
- Polyamide lining
- Polyurethane insole
- Steel toe cap
- Steel, puncture-resistant sole
- Purofort™ outsole
- Suitable for use in farming industry
Spezifikation
Green | |
Polyamide | |
Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole. | |
Steel | |
EN ISO 20345 : 2011 |
Agriculture/Farming | |
41 | |
Purofort™ | |
900 g per boot in size 9.5 | |
Safety Boot |