Learn More
STEMCELL Technologies Purmorphamine
Beschreibung
- Purmorphamine is a tri-substituted purine derivative that activates the Hedgehog pathway by directly binding to and activating the Hedgehog receptor Smoothened (EC50 = 1 μM)
- (Sinha and Chen)
- Promotes differentiation of ventral spinal progenitors and motor neurons from human pluripotent stem cells (Hu and Zhang, Karumbayaram et al.), Li et al.)
- Promotes differentiation of osteoblasts from human and mouse mesenchymal cells (Beloti et al.), Wu et al. 2002, Wu et al.) 2004)
- Inhibits differentiation and maturation of adipocytes from human mesenchymal cells (Fontaine et al.)
Spezifikation
Spezifikation
| CAS | 483367-10-8 |
| Menge | 5 mg |
| Summenformel | C31H32N6O2 |
| Reinheit | ≥ 98% |