missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18128-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.Spezifikation
PURB | |
Polyclonal | |
Unconjugated | |
PURB | |
PURB, PURBETA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19291, 498407, 5814 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35295, Q68A21, Q96QR8 | |
Purb | |
PURB Fusion Protein Ag12705 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Liquid |