Learn More
PTPN13/PTPL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
237.00 CHF - 551.00 CHF
Spezifikation
| Antigen | PTPN13/PTPL1 |
|---|---|
| Verdünnung | Western Blot 1:2000 - 1:6000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Anwendungen | Western Blot, Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18693032
|
Novus Biologicals
NBP2-94207-0.02ml |
0.02 mL |
237.00 CHF
0.02 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18623352
|
Novus Biologicals
NBP2-94207-0.1ml |
0.1 mL |
551.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
PTPN13/PTPL1 Polyclonal antibody specifically detects PTPN13/PTPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpezifikation
| PTPN13/PTPL1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 5783 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:6000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1EDKFZp686J1497, PTP-BASEC 3.1.3.48, PTP-E1, PTPL1nonreceptor type 13, tyrosine-protein phosphatase non-receptor type 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-500 of human PTPN13 (NP_542416.1). QDENYFKDILSDNSGREDSENTFSPYQFKTSGPEKKPIPGIDVLSKKKIWASSMDLLCTADRDFSSGETATYRRCHPEAVTVRTSTTPRKKEARYSDGSIALDIFGPQKMDPIYHTRELPTSSAISSALDRIRERQKKLQVLREAMNVEEPVRRYKTYHGDVFSTSSESPSIISSESDFRQVRRSEASKRFESSSGLPGVDETLSQGQSQRPSRQYETPFEGNLINQEIMLKRQEEELMQLQAKMALRQSR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |