missing translation for 'onlineSavingsMsg'
Learn More

PSME2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen PSME2
Konzentration 0.33 mg/mL
Anwendungen Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16850904
Unterlagen ansehen Aktionsdetails
Proteintech
12937-2-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16840904
Unterlagen ansehen Aktionsdetails
Proteintech
12937-2-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Proteolytic degradation is critical to the maintece of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (approximately 31), barrel-shaped molecular machine with an apparent molecular weight of approximately 2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman′s syndrome, and Liddle syndrome.
Spezifikation

Spezifikation

PSME2
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P97372, Q63798, Q9UL46
19188, 29614, 5721
PSME2 Fusion Protein Ag3986
Primary
-20°C
PSME2
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PA28B, PA28beta, PSME2, REG beta, REGbeta
Psme2
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications