Learn More
PSMD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544971
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QVRTARHNII CLQNDHKAVM KQVEEALHQL HARDKEKQAR DMAEAHKEAM Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
PSMD9 Polyclonal antibody specifically detects PSMD9 in Human samples. It is validated for Western Blot.Spezifikation
PSMD9 | |
Polyclonal | |
Unconjugated | |
PSMD9 | |
1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5715 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00233 | |
PSMD9 | |
Synthetic peptide directed towards the middle region of human PSMD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |