missing translation for 'onlineSavingsMsg'
Learn More

PSMD9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA5121663

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 17713064

  • 200.00 CHF / 20 Mikroliter
Verfügbar ab: 18-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
Spezifikation

Spezifikation

PSMD9
Polyclonal
Unconjugated
PSMD9
1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1
Rabbit
Antigen affinity chromatography
RUO
5715
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 50% glycerol and 0.05% ProClin 300
O00233
PSMD9
Recombinant protein Proteasome 26S Subunit, Non ATPase 9. The antigen corresponds to amino acid range 2-223 of the target protein.
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote