Learn More
PSMD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA592418
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: MSDEEARQSG GSSQAGVVTV SDVQELMRRK EEIEAQIKAN YDVLESQKGI GMNEPLVDCE GYPRSDVDLY QVRTARHNII CLQNDHKAVM KQVEEALHQL HARDKEKQAR DMAEAHKEAM SRKLGQSESQ GPPRAFAKVN SISPGSPASI AGLQVDDEIV EFGSVNTQNF QSLHNIGSVV QHSEGKPLNV TVIRRGEKHQ LRLVPTRWAG KGLLGCNIIP LQR; Positive Samples: H460, 22Rv1, Jurkat, A-549
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.Spezifikation
PSMD9 | |
Polyclonal | |
Unconjugated | |
PSMD9 | |
1500011J20Rik, 2610202L11Rik, 26S proteasome non-ATPase regulatory subunit 9, 26S proteasome regulatory subunit p27, Bridge, Bridge-1, homolog of rat Bridge 1, hypothetical protein LOC513315, MGC8644, P27, proteasome (prosome, macropain) 26S subunit, non-ATPase, 9, proteasome 26S non-ATPase regulatory subunit 9, proteasome 26S non-ATPase subunit 9, proteasome 26S subunit, non-ATPase 9, Psmd9, Rpn4, Transactivating protein Bridge-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
161475, 5715, 67151 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
O00233, Q9CR00, Q9WTV5 | |
PSMD9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-223 of human PSMD9 (NP_0028042) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |