Learn More
PSMD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560652
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ASIAGLQVDD EIVEFGSVNT QNFQSLHNIG SVVQHSEGKP LNVTVIRRGE KHQLRLVPTR WAGKGLLGCN IIPLQR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.Spezifikation
PSMD9 | |
Polyclonal | |
Unconjugated | |
PSMD9 | |
1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5715 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00233 | |
PSMD9 | |
Recombinant protein corresponding to Human PSMD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only