missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18109-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peripherin-2 (PRPH2), also known as retinal degeneration slow protein (RDS), is a photoreceptor-specific tetraspanin protein implicated in outer segment disk morphogenesis. It may function as an adhesion molecule involved in stabilization and compaction of outer segment disks or in the maintece of the curvature of the rim. Mutations in peripherin-2 are responsible for various retinal degenerative diseases including autosomal domit retinitis pigmentosa (ADRP).Spezifikation
PRPH2 | |
Polyclonal | |
Unconjugated | |
PRPH2 | |
AOFMD, AVMD, Peripherin 2, Peripherin2, PRPH, PRPH2, rd2, RDS, RP7, Tetraspanin 22, Tspan 22, TSPAN22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19133, 25534, 5961 | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15499, P17438, P23942 | |
Prph2 | |
PRPH2 Fusion Protein Ag12555 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |