Learn More
R&D Systems™ Proteome Profiler Human Ubiquitin Array
Beschreibung
The Proteome Profiler Human Ubiquitin Array contains 4 membranes-each spotted in duplicate with 49 different antibodies to ubiquitin target proteins.
Spezifikation
Spezifikation
| Inhalt und Lagerung | 4 Array Membranes, 4-Well Multi-Dish, Array Buffers, Wash Buffers, Detection Antibody Cocktail, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol. Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Produktlinie | R&D Systems |
| Menge | 1 Kit |