Learn More
R&D Systems™ Proteome Profiler Human Protease/Protease Inhibitor Array
Beschreibung
The Proteome Profiler Human Protease/Protease Inhibitor Array contains 4 membranes-2 spotted in duplicate with antibodies against 35 different proteases and 2 spotted in duplicate with antibodies against 32 different protease inhibitors.
Spezifikation
Spezifikation
| Inhalt und Lagerung | 4 Array Membranes (2 of Proteases and 2 Protease Inhibitors), 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Human Protease Array, Detection Antibody Cocktail, Human Protease Inhibitor Array, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol. Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Produktlinie | R&D Systems |
| Menge | 1 Kit |