Learn More
Cytiva™ Protein Select™ Resin
Beschreibung
- Infrared (IR) turbine rotor reflection ensures accurate and repeatable flow measurements, ideal for precision-critical applications
- Robust and stable design with four screw gaps allows for secure and easy mounting in various system configurations
- Constructed from USP Class VI PVDF material, offering excellent chemical resistance and compatibility with aggressive liquids
- Supports gamma sterilization up to 50kGy, making it suitable for use in sterile environments and medical-grade processes
- Delivers high-resolution square wave output for reliable signal processing and integration with control systems
- Tubes can be sterilized up to 150°C (302°F), enabling safe use in high-temperature cleaning and sterilization protocols
- Designed to reduce downtime and prevent cross-contamination, enhancing operational efficiency and product safety
- Each sensor is pre-calibrated to achieve accuracy within <1%, minimizing setup time and ensuring consistent performance
Spezifikation
Spezifikation
| Typ | Durchflussmesser |
| Genauigkeit | ± 1 % |
| Luftstromrate | 0.03 to 2 L/min. |
| Gehäusematerial | PVDF |
| Float Material | PVDF |
| Gastyp | Alle |
| Nenndruck | 362 psig |
| Enthält | 1/8 NPT Tubeholder System |