missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein O-Fucosyltransferase 1/POFUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
469.00 CHF - 703.00 CHF
Spezifikation
| Antigen | Protein O-Fucosyltransferase 1/POFUT1 |
|---|---|
| Verdünnung | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18627085
|
Novus Biologicals
NBP2-49136-25ul |
25 μL |
469.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18672056
|
Novus Biologicals
NBP2-49136 |
0.1 mL |
703.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Protein O-Fucosyltransferase 1/POFUT1 Polyclonal antibody specifically detects Protein O-Fucosyltransferase 1/POFUT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| Protein O-Fucosyltransferase 1/POFUT1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 23509 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FUT12EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 1, KIAA0180MGC2482, o-fucosyltransferase protein, O-Fuc-T, O-FucT-1, O-FUT, Peptide-O-fucosyltransferase 1, protein O-fucosyltransferase 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LPDLKEIQRAVKLWVRSLDAQSVYVATDSESYVPELQQLFKGKVKVVSLKPEVAQVDLYILGQADHFIGNCVSSFTAFVKRERDLQGRPSSFFGMD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts