missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ 3R SAX 3 μm HPLC Columns

Artikelnummer. 17817903 Alle ansehen Thermo Scientific Produkte
Click to view available options
Länge (metrisch):
50 mm
100 mm
Durchmesser (metrisch):
2 mm
4 mm
Flussrate:
0,1 bis 0,2 ml/min
0,3 bis 0,5 ml/min
Packungsgröße:
1 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17817903

Marke: Thermo Scientific™ 43203054068

Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Get the performance you need with ProPac™ 3R SAX HPLC columns. Suitable for charge variant separations and recommended for therapeutic proteins, mAbs, ADCs, and AAVs.

Use Thermo Scientific™ ProPac™ 3R SAX 3 μm HPLC columns to provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins with acidic pI values, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), biosimilar proteins, and adeno-associated viruses (AAVs).

Designed with bio-inert materials to reduce secondary interactions, our ProPac 3R SAX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.

Suitable for use with mass spectrometry (MS).

Maximizing Reproducibility, Resolution, and Robustness

The ProPac 3R SAX 3 μm HPLC column is a strong anion-exchanger with a quaternary amine functional group, suitable for use in biomolecule applications. These specialized ion-exchange chromatography columns provide qualitative and quantitative evaluation of charge variant analysis to biopharmaceutical scientists who need reproducible resolving capabilities when identifying and quantifying charge variant profiles of proteins.

Our ProPac 3R SAX columns offer superior performance for characterization of proteins including monoclonal antibodies (mAbs), therapeutic proteins with acidic pI values, antibody drug conjugates (ADCs), biosimilar proteins, and adeno-associated viruses (AAV).

Column Features:

  • Ideal for characterization and quality control assessment of closely-related protein variants
  • Ability to separate full versus empty capsids for AAVs
  • The high sample loading capability, without the carryover, allows for easy method optimization to provide you with excellent sensitivity and performance over a broad range of pH's, temperatures, and mobile phase compositions
  • Compatible with mass spectrometry (MS)
  • High-efficiency separations, resulting in a better understanding of charge variants in your samples
  • Higher capacity than other columns based on nonporous particles
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing

We recommend our CX-1 pH gradient buffers when using these columns with LC-UV/LC-FLD.

TRUSTED_SUSTAINABILITY

Spezifikation

Säulenformat Analysesäule
Durchmesser (metrisch) 4 mm
Flussrate 0,3 bis 0,5 ml/min
Länge (metrisch) 50 mm
Max. Druck 4500 psi (310 bar)
Partikelgröße 3 μm
pH 2 bis 12
Porengröße Nicht porös
Temperatur Raumtemperatur bis 60 °C
Stationäre Phase Geschützt
Produktlinie ProPac
Verpackungsmaterial Monodisperse Polymer Resin
Säulentyp Ionenaustausch
Phase Strong Anion Exchange
Material PEEK Hardware
Verfahren Bio-LC
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt