missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pro-Brain Natriuretic Peptide Monoclonal Antibody (5B6)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA120622
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MA1-20622 detects proBNP in human samples.
In cardiac tissue brain natriuretic peptide (BNP) is synthesized as 134 amino acid precursor (prepro-BNP), which is cleaved by proteases to form a 26 aa 'signal' peptide and a 108 aa pro-BNP. Proteolytic digestion of pro-BNP results in formation of 76 aa amino-terminal NT-proBNP and biologically active 32 aa BNP hormone molecule. Both proBNP and NTproBNP circulate in human plasma and have been proposed as markers for early diagnosis of left ventricular dysfunction as well as prognostic markers of possible cardiac complications at patients with heart failure.
Spezifikation
| pro-Brain Natriuretic Peptide | |
| Monoclonal | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P16860 | |
| NPPB | |
| Synthetic N-terminal pro brain natriuretic peptide (NT-proBNP), corresponding to amino acid residues 1-12. The peptides used for immunization were conjugated with carrier protein. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 5B6 | |
| Unconjugated | |
| NPPB | |
| 5 kDa cardiac natriuretic peptide; AA408272; BNF; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; NTproBNP; pro-BNP; type-B natriuretic factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 4879 | |
| 4°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur