missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRKCG (PKC gamma), GST Tag Recombinant Protein

Artikelnummer. p-7180469
Click to view available options
Menge:
1 mg
10 μg
100 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17717517

Marke: Invitrogen™ A41917

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Active The gene accession number for PRKCG (PKC gamma) is NP_002730.1.

The PKC family of serine/threonine kinases, including PRKCG (PKC gamma), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. PKC gamma is one of the PKC family members. This protein kinase is expressed solely in the brain and spinal cord and its localization is restricted to neurons. It has been demonstrated that several neuronal functions, including long term potentiation (LTP) and long term depression (LTD), specifically require this kinase. Knockout studies in mice also suggest that this kinase may be involved in neuropathic pain development. Defects in this protein have been associated with neurodegenerative disorder spinocerebellar ataxia-14 (SCA14).
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P05129
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM tris HCl with 20% glycerol, 100mM NaCl, 15mM glutathione, 5mM DTT and no preservative; pH 8
Gen-ID (Entrez) 5582
Molekulargewicht 112.3 kDa
Name Human PRKCG (PKC gamma), GST Tag
pH-Bereich 8
Reinigungsverfahren Purified
Menge 10 μg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias PKC; Pkcc; PKCG; PKCgamma; PKC-gamma; PKCI; Prkc; Prkcc; PRKCG; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma; RATPKCI; SCA14
Gebräuchliche Bezeichnung PKC gamma
Gensymbol PRKCG
Kinase-Familie PKC Kinase Family
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung GST-tag
Sequenz Full length
Inhalt und Lagerung -80°C, Avoid Freeze/Thaw Cycles
Versandbedingung Dry Ice
Expressionssystem Insect cells
Proteinfamilie Kinases & Inhibitors
Proteinform Full Length, Recombinant, Active
Form Liquid
Protein-Subtyp Serine/Threonine Kinases
Forschungskategorie Signal Transduction
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt