Learn More
Tocris Bioscience™ Pregnenolone sulfate sodium salt
GREENER_CHOICE
Artikelnummer. 18724049
Click to view available options
Menge:
50 mg
Packungsgröße:
50 Milligramm
Artikelnummer. 18724049
Marke: Tocris Bioscience™ 5376/50
om dit product te kopen Registreer vandaag om een webaccount aan te maken
NMDA potentiator; also GABAA inhibitor and TRPM3 activator
Pregnenolone sulfate sodium salt is a highly potent neurosteroid with multiple actions. Enhances intracellular Ca2+ upon NMDA receptor-mediated synaptic activity (EC50 ∽ 2 pM) and potentiates NMDA responses. Inhibitor at GABAA receptors (IC50 values are 0.4 to >300 μM, depending on specific subunit conformation). Activates CREB phosphorylation, augments LTP and cognitive behavior in rat memory models. Also activates TRPM3 channels.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 50 mg
Produkttyp Pregnenolone sulfate sodium salt
Molekulargewicht 418.52
Hemmstoffe Pregnenolone sulfate sodium salt
Nur für Forschungszwecke