missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAS40 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21097-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can indusce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.Spezifikation
PRAS40 | |
Polyclonal | |
Unconjugated | |
AKT1S1 | |
AKT1S1, AKTS1, Lobe, PRAS40, Proline rich AKT1 substrate 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84335 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96B36 | |
AKT1S1 | |
PRAS40 Fusion Protein Ag15355 | |
150 μL | |
Primary | |
Human | |
Liquid |