missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPP1R3C (Human) Recombinant Protein
Human PPP1R3C full-length ORF ( AAH12625, 1 a.a. - 317 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00005507-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MSCTRMIQVLDPRPLTSSVMPVDVAMRLCLAHSPPVKSFLGPYDEFQRRHFVNKLKPLKSCLNIKHKAKSQNDWKCSHNQAKKRVVFADSKGLSLTAIHVFSDLPEEPAWDLQFDLLDLNDISSALKHHEEKNLILDFPQPSTDYLSFRSHFQKNFVCLENCSLQERTVTGTVKVKNVSFEKKVQIRITFDSWKNYTDVDCVYMKNVYGGTDSDTFSFAIDLPPVIPTEQKIEFCISYHANGQVFWDNNDGQNYRIVHVQWKPDGVQTQMAPQDCAFHQTSPKTELESTIFGSPRLASGLFPEWQSWGRMENLASYR
Spezifikation
AAH12625 | |
protein phosphatase 1, regulatory (inhibitor) subunit 3C | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPP1R3C | |
GST |
5507 | |
Wheat germ expression system | |
25 μg | |
PPP1R5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |