missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R1C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539369
Additional Details : Gewicht : 0.00900kg
Beschreibung
Protein phosphatase-1 (PP1) is a major serine/threonine phosphatase that regulates a variety of cellular functions. PP1 consists of a catalytic subunit (see PPP1CA; MIM 176875) and regulatory subunits that determine the subcellular localization of PP1 or regulate its function. PPP1R1C belongs to a group of PP1 inhibitory subunits that are themselves regulated by phosphorylation (Wang et al., 2008 [PubMed 18310074]).[supplied by OMIM, Feb 2010].Spezifikation
PPP1R1C | |
Polyclonal | |
Unconjugated | |
PPP1R1C | |
PPP1R1C | |
A synthetic peptide derived from the internal region of human PPP1R1C | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WVI7, Q8BKK4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151242, 75276 | |
-20°C | |
Liquid |